Single-component explosives solutions
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31452 31453 31498 31600 31601 31602 31657 31658 31659 31660 31661 31662 31663 31664 31665 31666 31668 31669 31670 31671 33901
REST31452EA 52 EUR InStock
REST31452 REST31453 REST31498 REST31600 REST31601 REST31602 REST31657 REST31658 REST31659 REST31660 REST31661 REST31662 REST31663 REST31664 REST31665 REST31666 REST31668 REST31669 REST31670 REST31671 REST33901
Single-component explosives solutions
Standards Organische Standards
Single-component explosives solutions supports US department of defense base closures and remediation.
  • Mixtures and singles supporting LC US EPA method 8330
  • Mixtures and singles supporting GC-ECD US EPA method 8095
  • Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge