ASTM D2887-12 calibration standard (20 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31674
REST31674EA 116.5 EUR InStock
REST31674
ASTM D2887-12 calibration standard (20 components)
Standards Mineralölindustrie-Standards
Designed to serve as a column resolution text mixture, as well as a calibration mixture for retention time (RT) versus boiling point (bp) calibration.
The mix consists of, (C5) n-pentane, (C6) n-hexane, (C7) n-heptane, (C8) n-octane, (C9) n-nonane, (C10) n-decane, (C11) n-undecane, (C12) n-dodecane, (C14) n-tetradecane, (C15) n-pentadecane, (C16) n-hexadecane, (C17) n-heptadecane, (C18) n-octadecane, (C20) n-eicosane, (C24) n-tetracosane, (C28) n-octacosane, (C32) n-dotriacontane, (C36) n-hexatriacontane, (C40) n-tetracontane, (C44) n-tetratetracontane.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge