System performance test standard mixture of n-Alkanes (16 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31678
REST31678EA 108.5 EUR InStock
REST31678
System performance test standard mixture of n-Alkanes (16 components)
Standards Umweltanalyse-Standards
This standard mixture cobtains: (C10) n-Decane, (C12) n-Dodecane, (C14) n-Tetradecane, (C16) n-Hexadecane, (C18) n-Octadecane, (C20) n-Eicosane, (C22) n-Docosane, (C24) n-Tetracosane, (C26) n-Hexacosane, (C28) n-Octacosane, (C30) n-Triacontane, (C32) n-Dotriacontane, (C34) n-Tetratriacontane, (C36) n-Hexatriacontane, (C38) n-Octatriacontane and (C40) n-Tetracontane.
Shelf life 6 to 96 months.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge