Extraction solvent stock solution #2 (2 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31680
REST31680EA 114.5 EUR InStock
REST31680
Extraction solvent stock solution #2 (2 components)
Standards Umweltanalyse-Standards
This extraction solvent stock solution contains: n-decane (C10) (20 µl/l), n-tetracontane (C40) (20 mg/l).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge