CCME PHC calibration mix (3 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31684
REST31684EA 50 EUR InStock
REST31684
CCME PHC calibration mix (3 components)
Standards Organische Standards
This calibration mix consist of three components: (C10) n-Decane, (C16) n-Hexadecane and (C34) n-Tetratriacontane.
  • Meets CCME 2001 petroleum hydrocarbons in soil method—Tier 1
  • Primary reference calibration standards for quantification of four fractions
Shelf life 6 to 36 months.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge