Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Fragrance materials test mix (12 components)

Lieferant: RESTEK

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


31807
REST31807EA 274 EUR InStock
REST31807
Fragrance materials test mix (12 components)
Standards Organische Standards
This fragrance materials test mix idela to performance evaluation for essential oils and fragrance chemicals.

  • Performance evaluation for essential oils and fragrance chemicals
  • System suitability mixture for GC systems and analytical columns
  • Convenient 0,5 ml quantity for easy dilution to recommended working solution

This specially formulated reference standard kit exceeds IFRA purity requirements and contains all 24 EU-defined fragrance allergens detectable by GC-MS, along with seven additional potential allergens. When analyzed on a Restek Rxi-17 column following the IFRA method, all required compounds and isomers resolve clearly with no coelutions. This time-saving mix is also incredibly robust with a shelf life of over one and a half years, so you can stock an already-made standard instead of spending the time to create a new batch for every analysis.

It contain 12 components: Benzoic acid (1,0%), Benzyl salicylate (36,2%), 1,8-Cineole (Eucalyptol) (0,5%), trans Cinnamaldehyde (0,5%), Cinnamyl acetate (0,3%), Cinnamyl alcohol 0,3%, Ethyl butyrate (36,2%), Geraniol (0,6%), Hydroxycitronellal (3,7-Dimethyl-7-hydroxyoctanal) (5,0%), d-Limonene (20,0%), Thymol (0,3%) and Vanilliv (0,1%).

Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge