Canadian drinking water phenoxyacid herbicides mix (11 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31868
REST31868EA 98 EUR InStock
REST31868
Canadian drinking water phenoxyacid herbicides mix (11 components)
Standards Umweltanalyse-Standards
Consists of 11 components, those are bromoxynil, 2,4-d, dicamba, 2,4-dichlorophenol, diclofop methyl, dinoseb, pentachlorophenol, picloram, 2,4,5-t, 2,3,4,6-tetrachlorophenol and 2,4,6-trichlorophenol.
Zertifizierungen: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge