Atlantic RBCA VPH mix (12 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31871
REST31871EA 51 EUR InStock
REST31871
Atlantic RBCA VPH mix (12 components)
Standards Umweltanalyse-Standards
Contains: n-hexane, C6, n-heptane, C7, n-octane, C8, n-decane, C10, benzene, ethylbenzene, 1-methyl-3-ethylbenzene, toluene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, o-xylene, p-xylene.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge