EPA method 8310 PAH mixture
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31874
REST31874EA 139.5 EUR InStock
REST31874
EPA method 8310 PAH mixture
Standards Umweltanalyse-Standards
EPA method 8310 PAH mixturecontain 18 components.
Matrix: Acetonitrile:toluene (92:8)
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge