Sorbitol standard mix (2 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31893
REST31893EA 0 EUR InStock
REST31893
Sorbitol standard mix (2 components)
Standards Organische Standards
Testing of glycerin for diethylene glycol with diethylene glycol (DEG) and ethylene glycol limit standards.
This guidance emphasizes the importance of screening raw material for the presence of diethylene glycol. Under cGMPs, drug manufacturers, not just glycerin manufacturers, must test glycerin prior to use to prevent DEG contamination in finished products. The FDA has worked extensively with the USP to modify the glycerin monograph, and these standards support the revised USP method.
Consists of diethylene glycol and ethylene glycol.
Zertifizierungen: Meet FDA guidance for industry.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge