Herbicide mix #1 (7 components), free acid form
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32054
REST32054EA 79.5 EUR InStock
REST32054
Herbicide mix #1 (7 components), free acid form
Standards Organische Standards
This mix contains seven components.
Components are 2,4-D, 2,4-DB, 2,4,5-T, 2,4,5-TP, dicamba, dichlorprop, dinoseb.
Contains: 2,4-D (94-75-7); 2,4-DB (94-82-6); 2,4,5-T (93-76-5); 2,4,5-TP (Silvex) (93-72-1); Dicamba (1918-00-9); Dichlorprop (120-36-5); Dinoseb (88-85-7)
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge