Herbicide mix #1 (7 components), derivatised form
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32055
REST32055EA 91 EUR InStock
REST32055
Herbicide mix #1 (7 components), derivatised form
Standards Organische Standards
This mix contains seven components.
Components are 2,4-D methyl ester, 2,4-DB methyl ester, 2,4,5-T methyl ester, 2,4,5-TP methyl ester, dicamba methyl ester, dichlorprop methyl ester, dinoseb methyl ether.
Contains: 2,4-D methyl ester (1928-38-7); 2,4-DB methyl ester (18625-12-2); 2,4,5-T methyl ester (1928-37-6); 2,4,5-TP methyl ester (Silvex) (4841-20-7); Dicamba methyl ester (6597-78-0); Dichlorprop methyl ester (57153-17-0); Dinoseb methyl ether (6099-79-2)
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge