Herbicide mix #4 (8 components), free acid form
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32061
REST32061EA 64 EUR InStock
REST32061
Herbicide mix #4 (8 components), free acid form
Standards Organische Standards
This mix contains eight components.
Components are acifluorfen, bentazon, chloramben, DCPA diacid (tetrachloroterephthalic acid), 3,5-dichlorobenzoic acid, 4-nitrophenol, pentachlorophenol, picloram.
Contains: Acifluorfen (50594-66-6); Bentazon (25057-89-0); Chloramben (133-90-4); DCPA diacid (tetrachloroterephthalic acid) (2136-79-0); 3,5-Dichlorobenzoic acid (51-36-5); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Picloram (1918-02-1)
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge