PCB congener mix, method 8082A (19 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32416
REST32416EA 253.5 EUR InStock
REST32416
PCB congener mix, method 8082A (19 components)
Standards Organische Standards
Contains 19 components, those are 2-chlorobiphenyl , 2,3-dichlorobiphenyl , 2,2',5-trichlorobiphenyl , 2,4',5-trichlorobiphenyl, 2,2',3,5'-tetrachlorobiphenyl, 2,2',5,5'-tetrachlorobiphenyl, 2,3',4,4'-tetrachlorobiphenyl, 2,2',3,4,5'-pentachlorobiphenyl, 2,2',4,5,5'-pentachlorobiphenyl, 2,3,3',4',6-pentachlorobiphenyl, 2,2',3,4,4',5'-hexachlorobiphenyl, 2,2',3,4,5,5'-hexachlorobiphenyl, 2,2',3,5,5',6-hexachlorobiphenyl, 2,2',4,4',5,5'-hexachlorobiphenyl, 2,2',3,3',4,4',5-heptachlorobiphenyl, 2,2',3,4,4',5,5'-heptachlorobiphenyl, 2,2',3,4,4',5',6-heptachlorobiphenyl, 2,2',3,4',5,5',6-heptachlorobiphenyl and 2,2',3,3',4,4',5,5',6-nonachlorobiphenyl.
Zertifizierungen: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge