GC multi-residue pesticide standard #8 (24 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
32570
REST32570EA 331 EUR InStock
REST32570
GC multi-residue pesticide standard #8 (24 components)
Standards Organische Standards
The multiresidue accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability
The multiresidue pesticide standard consist of 24 components bromfenvinfos-methyl, bromfenvinphos, bromophos ethyl, bromophos methyl, carbophenothion, chlorfenvinphos, chlorthiophos, coumaphos, edifenphos, ethion, fenamiphos, fenchlorphos (ronnel), fenthion, iodofenphos, leptophos, malathion, methacrifos, profenofos, prothiofos, sulphotepp, sulphrofos, terbufos, tetrachlorvinfos, tolclofos-methyl.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge