Fragrance allergen standards kit
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
33105
REST33105EA 587.5 EUR InStock
REST33105
Fragrance allergen standards kit
Standards Organische Standards
Ideal for GC-MS analysis following IFRA methodology.
  • Fully resolves 31 fragrance allergens with one analysis on an Rxi-17 column
  • Helps you meet EU requirements defined in the European Cosmetics Directive
  • Exceeds purity requirements outlined in IFRA method
  • All components included at 400 ppm to allow dilution for calibration curves and use with different solvents
This specially formulated reference standard kit exceeds IFRA purity requirements and contains all 24 EU-defined fragrance allergens detectable by GC-MS, along with seven additional potential allergens. When analyzed on a Restek Rxi-17 column following the IFRA method, all required compounds and isomers resolve clearly with no coelutions. This time-saving mix is also incredibly robust with a shelf life of over one and a half years, so you can stock an already-made standard instead of spending the time to create a new batch for every analysis.
Whether you manufacture fragrances in Europe, sell to European consumers, or simply wish to test for allergens in your products, choose the reference standard mix that helps you follow IFRA methodology in one convenient, long-lasting formulation.
Of those 26, 24 are volatile chemicals that can be determined through GC-MS analysis, and version 3 of the method outlined by the International Fragrance Association (IFRA) does just that.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge