LC reversed phase test mix #1 (4 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
35005
REST35005EA 62 EUR InStock
REST35005
LC reversed phase test mix #1 (4 components)
Standards Organische Standards
Routine analysis using this mix can assist in determining the need to perform column and/or system maintenance.
The LC reversed phase test mix #1 consists of four components, they are benzene (3,00 mg/ml), biphenyl (0.06 mg/ml), naphthalene (0,50 mg/ml) and uracil (0,02 mg/ml).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge