FAME #16 mix
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
35022
REST35022EA 118 EUR InStock
REST35022
FAME #16 mix
Standards Lebensmittel-Standards
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like corn, poppy seed, cotton seed, soybean, walnut, safflower, sunflower, rice, bran, and sesame oil.
This mix contains six components such as C16:00 methyl palmitate (6%), C18:00 methyl stearate (3%), C18:01 methyl oleate (35%), C18:02 methyl linoleate (50%), C18:03 methyl linolenate (3%), C20:00 methyl arachidate (3%).
% represents weight/weight% basis (minimum 50 mg/ampoule).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge