Free fatty acids test standard (6 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
35272
REST35272EA 64 EUR InStock
REST35272
Free fatty acids test standard (6 components)
Standards Organische Standards
The free fatty acids test standard consists of six components they are (C4:0) butyric acid, acetic acid, isobutyric acid, isovaleric acid, propionic acid and valeric acid.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge