Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid mit 1% tert-Butylchlordimethylsilan

Lieferant: RESTEK

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

35610 35608
REST35610EA 373 EUR InStock
REST35610 REST35608
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid mit 1% tert-Butylchlordimethylsilan
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid mit 1% tert-Butylchlordimethylsilan
Silylation derivatization reagent.

  • Replace active hydrogen, reducing polarity and making the compounds more volatile
  • Increase stability of derivatives

Silylation is the most widely used derivatization procedure for sample analysis by GC. In silylation, an active hydrogen is replaced by an alkylsilyl group such as trimethylsilyl (TMS) or tert-butyldimethylsilyl (tert-BDMS). Silyl derivatives are more volatile, less polar, and more thermally stable. As a result, GC separation is improved and detection is enhanced.

Both TMS and tert-BDMS reagents are suitable for a wide variety of compounds and can be used for many GC applications. (Note that silylation reagents are generally moisture-sensitive and must be sealed to prevent deactivation.)


Formel: C₉H₁₈F₃NOSi
MW: 241.33 g/mol
MDL Number: MFCD00009671
CAS-Nummer: 77377-52-7
UN: 2924
ADR: 3.8 II
Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge