Residual solvents - Class 1 (5 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36279
REST36279EA 0 EUR InStock
REST36279
Residual solvents - Class 1 (5 components)
Standards Organische Standards
The solvents consists of five components, they are benzene (10 mg/ml), carbon tetrachloride (20 mg/ml), 1,2-dichloroethane 25 mg/ml, 1,1-dichloroethene (40 mg/ml), and 1,1,1-trichloroethane (50 mg/ml).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge