Pharmaceutical residual solvents, USP <467> singles
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
36403 36281 36282 36283 36285 36286 36287 36288 36289 36290 36291 36292 36293 36294 36295 36296 36297 36298 36299 36400 36401 36402 36404 36405 36406 36407 36408 36409 36410 36411 36412 36413 36414 36415 36416 36284
REST36403EA 45 EUR InStock
REST36403 REST36281 REST36282 REST36283 REST36285 REST36286 REST36287 REST36288 REST36289 REST36290 REST36291 REST36292 REST36293 REST36294 REST36295 REST36296 REST36297 REST36298 REST36299 REST36400 REST36401 REST36402 REST36404 REST36405 REST36406 REST36407 REST36408 REST36409 REST36410 REST36411 REST36412 REST36413 REST36414 REST36415 REST36416 REST36284
Pharmaceutical residual solvents, USP <467> singles
Standards Organische Standards
The United States Pharmacopeia (USP)<467> residual solvents is a widely used compendial method intended for identifying and quantifying residual solvents in drug substances, drug products, and excipients.
In an attempt to better mirror the International Conference on Harmonization (ICH)guidelines, the USP has adopted a more comprehensive methodology in residual solvent testing-the current USP30/NF25. The ICH publishes a guideline (Q3C) listing the acceptable amounts of solvent residues that can be present. In the ICH guideline, residual solvents are summarised by class according to their toxicity.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge