36403
36281
36282
36283
36285
36286
36287
36288
36289
36290
36291
36292
36293
36294
36295
36296
36297
36298
36299
36400
36401
36402
36404
36405
36406
36407
36408
36409
36410
36411
36412
36413
36414
36415
36416
36284
REST36403EA
45
EUR
InStock
REST36403
REST36281
REST36282
REST36283
REST36285
REST36286
REST36287
REST36288
REST36289
REST36290
REST36291
REST36292
REST36293
REST36294
REST36295
REST36296
REST36297
REST36298
REST36299
REST36400
REST36401
REST36402
REST36404
REST36405
REST36406
REST36407
REST36408
REST36409
REST36410
REST36411
REST36412
REST36413
REST36414
REST36415
REST36416
REST36284
Pharmaceutical residual solvents, USP <467> singles
Standards
Organische Standards
The United States Pharmacopeia (USP)<467> residual solvents is a widely used compendial method intended for identifying and quantifying residual solvents in drug substances, drug products, and excipients.
In an attempt to better mirror the International Conference on Harmonization (ICH)guidelines, the USP has adopted a more comprehensive methodology in residual solvent testing-the current USP30/NF25. The ICH publishes a guideline (Q3C) listing the acceptable amounts of solvent residues that can be present. In the ICH guideline, residual solvents are summarised by class according to their toxicity.