CarboBlack columns (packed)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
80127-810 80127-800 80127-800901 80127-820 80127-821 80127-821901 80127-830 80127-840 80127-845 80127-890900 80127-890901 80127-895 80451-800 80451-810 80451-820 80451-845901 80452-800 80453-800 80453-840901 80454-800 80454-810 80455-800 80456-800 80457-800 80457-810 80457-820 80457-830 80457-840 80457-894 80458-800 80458-810 80458-830 80459-800 80459-810 80460-800 80460-810 88012-800 88012-810 88012-895R 88013-800 88013-810 88013-895 80127-810901 80127-860 80451-840 80452-810 88012-893
REST80127-810EA 891 EUR InStock
REST80127-810 REST80127-800 REST80127-800901 REST80127-820 REST80127-821 REST80127-821901 REST80127-830 REST80127-840 REST80127-845 REST80127-890900 REST80127-890901 REST80127-895 REST80451-800 REST80451-810 REST80451-820 REST80451-845901 REST80452-800 REST80453-800 REST80453-840901 REST80454-800 REST80454-810 REST80455-800 REST80456-800 REST80457-800 REST80457-810 REST80457-820 REST80457-830 REST80457-840 REST80457-894 REST80458-800 REST80458-810 REST80458-830 REST80459-800 REST80459-810 REST80460-800 REST80460-810 REST88012-800 REST88012-810 REST88012-895R REST88013-800 REST88013-810 REST88013-895 REST80127-810901 REST80127-860 REST80451-840 REST80452-810 REST88012-893
CarboBlack columns (packed)
Chromatographiesäulen
Graphitised carbon black offers unique selectivity and very little adsorption for alcohol analyses. Two types of CarboBlack supports are available, CarboBlack B and CarboBlack C. CarboBlack B support, with its higher surface area, can hold up to a 10% loading of a non silicone liquid phase. CarboBlack C support can hold up to a 1% loading of a non silicone liquid phase.
  • CarboBlack packings are treated with KOH or picric acid for basic or acidic compounds
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge