HPLC columns, Ultra C8 (USP L7)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9103311 9103312 9103315 9103331 9103332 9103335 9103351 9103352 9103355 9103361 9103362 9103365 9103511 9103512 9103514 9103515 9103521 9103522 9103525 9103531 9103532 9103535 9103551 9103552 9103555 9103561 9103562 9103564 9103565 9103571 9103572 9103575 910350210 910350212 9103312-700 9103313-700 9103315-700 9103332-700 9103333-700 9103335-700 9103352-700 9103353-700 9103355-700 9103362-700 9103363-700 9103365-700 9103512-700 9103513-700 9103515-700 9103522-700 9103523-700 9103525-700 9103532-700 9103533-700 9103552-700 9103553-700 9103535-700 9103555-700 9103562-700 9103563-700 9103565-700 9103572-700 9103573-700 9103575-700 910331E 910333E 910335E 910336E 910351E 910352E 910353E 910355E 910356E 910357E
REST9103311EA 0 EUR InStock
REST9103311 REST9103312 REST9103315 REST9103331 REST9103332 REST9103335 REST9103351 REST9103352 REST9103355 REST9103361 REST9103362 REST9103365 REST9103511 REST9103512 REST9103514 REST9103515 REST9103521 REST9103522 REST9103525 REST9103531 REST9103532 REST9103535 REST9103551 REST9103552 REST9103555 REST9103561 REST9103562 REST9103564 REST9103565 REST9103571 REST9103572 REST9103575 REST910350210 REST910350212 REST9103312-700 REST9103313-700 REST9103315-700 REST9103332-700 REST9103333-700 REST9103335-700 REST9103352-700 REST9103353-700 REST9103355-700 REST9103362-700 REST9103363-700 REST9103365-700 REST9103512-700 REST9103513-700 REST9103515-700 REST9103522-700 REST9103523-700 REST9103525-700 REST9103532-700 REST9103533-700 REST9103552-700 REST9103553-700 REST9103535-700 REST9103555-700 REST9103562-700 REST9103563-700 REST9103565-700 REST9103572-700 REST9103573-700 REST9103575-700 REST910331E REST910333E REST910335E REST910336E REST910351E REST910352E REST910353E REST910355E REST910356E REST910357E
HPLC columns, Ultra C8 (USP L7)
Chromatographiesäulen
Ultra C8 is a conventional monomeric octylsilane column offering a shorter alkyl chain to provide less hydrophobic retention and improved basic peak shape. It is made of high-purity, type-B silica that minimises activity and creates high-density bonding. These columns are designed for selective and reliable HPLC applications.
  • Suitable for a wide range of compounds from acidic through slightly basic
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge