HPLC columns, Ultra IBD (USP L68)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9175312 9175315 9175331 9175332 9175333 9175335 9175351 9175352 9175355 9175361 9175362 9175363 9175365 9175511 9175512 9175513 9175515 9175521 9175522 9175523 9175525 9175531 9175532 9175533 9175535 9175551 9175552 9175553 9175555 9175561 9175562 9175563 9175565 9175571 9175572 9175573 9175575 917550210 917550212
REST9175312EA 686 EUR InStock
REST9175312 REST9175315 REST9175331 REST9175332 REST9175333 REST9175335 REST9175351 REST9175352 REST9175355 REST9175361 REST9175362 REST9175363 REST9175365 REST9175511 REST9175512 REST9175513 REST9175515 REST9175521 REST9175522 REST9175523 REST9175525 REST9175531 REST9175532 REST9175533 REST9175535 REST9175551 REST9175552 REST9175553 REST9175555 REST9175561 REST9175562 REST9175563 REST9175565 REST9175571 REST9175572 REST9175573 REST9175575 REST917550210 REST917550212
HPLC columns, Ultra IBD (USP L68)
Chromatographiesäulen
Ultra IBD is a polar-embedded column that acts as a strong hydrogen bonder. With a unique polar group this column is very retentive and selective for acids. It also provides symmetrical peak shape for strong bases. This columns are compatible with 100% aqueous mobile phases and can be used under reversed-phase or HILIC conditions to retain very polar, ionic compounds in highly organic mobile phases.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge