HPLC columns, Ultra Aqueous C18 (USP L1)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9178311 9178312 9178313 9178315 9178331 9178332 9178333 9178335 9178351 9178352 9178353 9178355 9178361 9178362 9178365 9178511 9178512 9178513 9178515 9178521 9178522 9178523 9178525 9178531 9178532 9178533 9178535 9178551 9178552 9178553 9178555 9178561 9178562 9178563 9178565 9178571 9178572 9178573 9178575 917850210 917850212
REST9178311EA 0 EUR InStock
REST9178311 REST9178312 REST9178313 REST9178315 REST9178331 REST9178332 REST9178333 REST9178335 REST9178351 REST9178352 REST9178353 REST9178355 REST9178361 REST9178362 REST9178365 REST9178511 REST9178512 REST9178513 REST9178515 REST9178521 REST9178522 REST9178523 REST9178525 REST9178531 REST9178532 REST9178533 REST9178535 REST9178551 REST9178552 REST9178553 REST9178555 REST9178561 REST9178562 REST9178563 REST9178565 REST9178571 REST9178572 REST9178573 REST9178575 REST917850210 REST917850212
HPLC columns, Ultra Aqueous C18 (USP L1)
Chromatographiesäulen
Ultra Aqueous C18 columns are made of high-purity, type-B silica that minimizes activity and creates high-density bonding, these columns are designed for selective and reliable HPLC applications. It is highly retentive and selective for reversed phase separations of polar analytes, and highly base deactivated. Compatible with highly aqueous (upto 100%) mobile phases.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge