HPLC columns, Viva C4 (USP L26)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9512511 9512512 9512515 9512521 9512522 9512525 9512531 9512532 9512535 9512551 9512552 9512555 9512561 9512562 9512565 9512571 9512572 9512575 951250210 951250212 9512512-700 9512513-700 9512515-700 951251E 9512522-700 9512523-700 9512525-700 951252E 9512532-700 9512533-700 9512535-700 951253E 9512552-700 9512553-700 9512555-700 951255E 9512562-700 9512563-700 9512565-700 951256E 9512572-700 9512573-700 9512575-700 951257E
REST9512511EA 658 EUR InStock
REST9512511 REST9512512 REST9512515 REST9512521 REST9512522 REST9512525 REST9512531 REST9512532 REST9512535 REST9512551 REST9512552 REST9512555 REST9512561 REST9512562 REST9512565 REST9512571 REST9512572 REST9512575 REST951250210 REST951250212 REST9512512-700 REST9512513-700 REST9512515-700 REST951251E REST9512522-700 REST9512523-700 REST9512525-700 REST951252E REST9512532-700 REST9512533-700 REST9512535-700 REST951253E REST9512552-700 REST9512553-700 REST9512555-700 REST951255E REST9512562-700 REST9512563-700 REST9512565-700 REST951256E REST9512572-700 REST9512573-700 REST9512575-700 REST951257E
HPLC columns, Viva C4 (USP L26)
Chromatographiesäulen
These columns are based on a wide pore material designed for optimal large-molecule separations. Base-deactivated, wide-pore packing exhibits excellent peak shape for a wide range of compounds. Less retention in reversed phase assays than Viva C18 or Viva C8.
  • Rugged, spherical particles with 300 Å pore size
  • High proportion of pore/surface area available to large molecules
  • Excellent for separating peptides or proteins
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge