Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

LC column, force biphenyl

Lieferant: Restek

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

9629565
REST9629565EA 760 EUR InStock
REST9629565
LC column, force biphenyl
Chromatographiesäulen
Force Biphenyl LC HPLC Columns are ideal for bioanalytical testing applications like drug and metabolite analyses.

  • Increased retention for dipolar, unsaturated, or conjugated solutes
  • Enhanced selectivity when used with methanolic mobile phase
  • Ideal for increasing sensitivity and selectivity in LC-MS analyses

Columns feature heightened selectivity and retention for compounds that are hard to resolve or elute early on C18 and other phenyl chemistries. Columns also limit ionization suppression and allow simple, MS-friendly mobile phases.

Force performance LC columns are long-lasting and reproducible, even under the stress of UHPLC. Force columns are fully scalable between 3 or 5 µm HPLC and 1,8 µm UHPLC, so you can easily transfer and optimize methods without extensive calculations.

The innovative biphenyl phase is the most popular LC stationary phase because it is particularly adept at separating compounds that are hard to resolve or that elute early on C18 and other phenyl chemistries. As a result, Biphenyl columns are extremely useful for bioanalytical testing applications like drug and metabolite analyses, especially those that require a mass spectrometer (MS). Increasing retention of early-eluting compounds can limit ionization suppression, and the heightened selectivity helps eliminate the need for complex mobile phases that are not well-suited for MS detection.

Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge