Buckets, Round, PP
Lieferant: RIXIUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
U-7S-1001-018K 7-1001-100 7-1001-010 7-1001-050 U-7S-1001-080K 7-5101-110 7-5101-055 7-1001-006 U-7S-1001-025K 7-1001-050-K 7-1001-100-K 7-1001-036-K 7-1001-300-K 7-1001-300 7-5101-180 U-7S-1001-020K U-7S-1001-030K 7-2001-180 7-1001-200 7-5101-600
RIXIU-7S-1001-018KEA 942 EUR InStock
RIXIU-7S-1001-018K RIXI7-1001-100 RIXI7-1001-010 RIXI7-1001-050 RIXIU-7S-1001-080K RIXI7-5101-110 RIXI7-5101-055 RIXI7-1001-006 RIXIU-7S-1001-025K RIXI7-1001-050-K RIXI7-1001-100-K RIXI7-1001-036-K RIXI7-1001-300-K RIXI7-1001-300 RIXI7-5101-180 RIXIU-7S-1001-020K RIXIU-7S-1001-030K RIXI7-2001-180 RIXI7-1001-200 RIXI7-5101-600
Buckets, Round, PP
Eimer
Buckets made of PP, white.
  • With or without UN approval
  • Very good chemical resistance
  • Very good temperature resistance
Universal multipurpose buckets with handle.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge