Solvent filter
Lieferant: ROBU GLASFILTER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
80052
ROBU80052EA 24 EUR InStock
ROBU80052
Solvent filter
Filter Flüssigkeits-Prozessfilter
This solvent filter is made of pure borosilicate glass.
  • Highly resistant to most chemicals such as solvents, acids and alkalis
  • Reduced particle emission, specially rinsed, with calibrated rim
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge