Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MAPK1 Rabbit Monoclonal Antibody (Unconjugated) [clone: 3G2]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

100-401-218
ROCK100-401-218EA 821 EUR InStock
ROCK100-401-218
Anti-MAPK1 Rabbit Monoclonal Antibody (Unconjugated) [clone: 3G2]
Antikörper
This antiserum was tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a predominant band approximately 42 kDa in size corresponding to p42 MAP Kinase (ERK2) by western blotting.

Recommended Dilutions: ELISA: 1:2000 to 1:10000; Western Blot: 1:1000-1:,2000; Immunoprecipitation: 5.0 µg/mg sample;

Type: Primary
Antigen: MAPK1
Clonality: Monoclonal
Clone: 3G2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge