Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-HIST1H3A Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

100-401-E81S
ROCK100-401-E81SEA 226 EUR InStock
ROCK100-401-E81S
Anti-HIST1H3A Rabbit Polyclonal Antibody
Antikörper
Histone H3 antibody has been tested for use in ELISA and western blot. For western blots expect a band of approximately 15.4 kDa in size corresponding tothe Histone 3 Protein. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:2000;

Type: Primary
Antigen: HIST1H3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Chicken, Dog, Monkey, Xenopus laevis, Arabidopsis Thaliana, Caenorhabditis Elegans, Drosophila

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge