Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IgG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

18-0217-32
ROCK18-0217-32EA 457 EUR InStock
ROCK18-0217-32
Anti-IgG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Fluorescent Mouse TrueBlot® Antibody Fluorescein is provided as a lyophilized powder. In order to conserve reagent, we recommend incubating the blots from minigels in sealed bags (removing as much air as possible) with minimal volume (2-3 mls).

Recommended Dilutions: Restore with deionised water (or equivalent); 100 µl.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: eB144
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge