Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-SIX3 Guinea Pig Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-201-A26S
ROCK200-201-A26SEA 226 EUR InStock
ROCK200-201-A26S
Anti-SIX3 Guinea Pig Polyclonal Antibody
Antikörper
This Protein A purified antibody has been tested for use in ELISA, immunofluorescence microscopy and western blotting.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 37 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Immunofluorescence: 1:200 - 1:1000; Western Blot: 1:1000; Immunochemistry: 1:200;

Type: Primary
Antigen: SIX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Guinea Pig
Isotype: IgG
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge