Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATM Mouse Polyclonal Antibody (Unconjugated) [clone: 10H11.E12]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-301-400
ROCK200-301-400EA 971 EUR InStock
ROCK200-301-400
Anti-ATM Mouse Polyclonal Antibody (Unconjugated) [clone: 10H11.E12]
Antikörper
Protein A Purified Mab anti-ATM has been tested by ELISA and western blotting against both the native and recombinant forms of the Protein. The antibody immunoprecipitates ATM from irradiated human and transfected mouse cells.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Immunofluorescence: 1:100 - 1:500; Western Blot: 1:200 - 1:2000;

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone: 10H11.E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge