Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-8-Hydroxy Guanine Mouse Polyclonal Antibody (Unconjugated) [clone: 15A3]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-301-A99
ROCK200-301-A99EA 841 EUR InStock
ROCK200-301-A99
Anti-8-Hydroxy Guanine Mouse Polyclonal Antibody (Unconjugated) [clone: 15A3]
Antikörper
This Protein G purified antibody has been tested for use in immunochemistry and ELISA.  Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:20000; Immunochemistry: 20 µg/ml;

Type: Primary
Antigen: 8-Hydroxy Guanine
Clonality: Polyclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: pan

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge