Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TRPC6 Mouse Monoclonal Antibody (Unconjugated) [clone: 3F2.H10.F2]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-301-B59S
ROCK200-301-B59SEA 226 EUR InStock
ROCK200-301-B59S
Anti-TRPC6 Mouse Monoclonal Antibody (Unconjugated) [clone: 3F2.H10.F2]
Antikörper
This monoclonal antibody is suitable for ELISA, immunochemistry and western blotting. Expect a band approximately 106 kDa in size corresponding to TRPC6 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 2.5 µg/ml;

Type: Primary
Antigen: TRPC6
Clonality: Monoclonal
Clone: 3F2.H10.F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge