Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GRIN2B Mouse Monoclonal Antibody (Unconjugated) [clone: S59-36]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-301-G23
ROCK200-301-G23EA 841 EUR InStock
ROCK200-301-G23
Anti-GRIN2B Mouse Monoclonal Antibody (Unconjugated) [clone: S59-36]
Antikörper
Anti-NR2B Antibody is suitable for use in WB, IHC and IP. Expect a band approximately ~166kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: GRIN2B
Clonality: Monoclonal
Clone: S59-36
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge