Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TLR3 Mouse Monoclonal Antibody (Unconjugated) [clone: 40C1285.6]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-301-I24
ROCK200-301-I24EA 939 EUR InStock
ROCK200-301-I24
Anti-TLR3 Mouse Monoclonal Antibody (Unconjugated) [clone: 40C1285.6]
Antikörper
Anti-TLR3 Antibody is suitable for use in Flow (CS), Flow (I), IF/ICC, IHC (p), IP and WB. Expect a band approximately 120kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Flow Cytometry: 2-4 µg/10^6 cells; Western Blot: 1-3 µg/ml; Immunochemistry: 10 ug/ml; Immunoprecipitation: 2 µg/10^6 cells

Type: Primary
Antigen: TLR3
Clonality: Monoclonal
Clone: 40C1285.6
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge