Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ErpD Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-401-C09
ROCK200-401-C09EA 821 EUR InStock
ROCK200-401-C09
Anti-ErpD Rabbit Polyclonal Antibody
Antikörper
Anti-ErpD antibody has been tested in Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect bands at ~37.5 kDa and ~65 kDa in size corresponding to ErpD/Arp37 by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:1000;

Type: Primary
Antigen: ErpD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge