Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-CDKN2A Rat Polyclonal Antibody (Unconjugated) [clone: 5.C3.1]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-501-891
ROCK200-501-891EA 841 EUR InStock
ROCK200-501-891
Anti-CDKN2A Rat Polyclonal Antibody (Unconjugated) [clone: 5.C3.1]
Antikörper
This antibody has been tested for use in ELISA, western blotting, immunofluorescence microscopy, and immunoprecipitation. Specific conditions for western blotting reactivity should be optimized by the end user.  Expect a band approximately 19 kDa in size.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Immunofluorescence: 1:500 - 1:2000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone: 5.C3.1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human, Hamster, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge