Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LAMP2 Rat Monoclonal Antibody (Unconjugated) [clone: GL2A7]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-501-G10
ROCK200-501-G10EA 841 EUR InStock
ROCK200-501-G10
Anti-LAMP2 Rat Monoclonal Antibody (Unconjugated) [clone: GL2A7]
Antikörper
Anti-LAMP2 Antibody is recommended for use in IF microscopy and IP. The antibody will label the presumptive lysosomes and late endosomes in cells that have been permeabilized with saponin.

Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: GL2A7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Rabbit, Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge