Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MBP Chicken Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

200-901-D81
ROCK200-901-D81EA 861 EUR InStock
ROCK200-901-D81
Anti-MBP Chicken Polyclonal Antibody
Antikörper
Anti-Myelin Basic Protein antibody is suitable for use in Western Blotting and IF. Expect a bands of approximately 18 kDa and 22 kDa in size corresponding to MBP Proteins in Western blots of Rat brain homogenate.

Recommended Dilutions: Immunofluorescence: 1:500; Western Blot: 1:5000

Type: Primary
Antigen: MBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Bovine, Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge