Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ESRP2 Mouse Monoclonal Antibody (Unconjugated) [clone: 22C5.H7.A6]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

210-301-C32S
ROCK210-301-C32SEA 226 EUR InStock
ROCK210-301-C32S
Anti-ESRP2 Mouse Monoclonal Antibody (Unconjugated) [clone: 22C5.H7.A6]
Antikörper
This Protein-A purified antibody has been tested for use western blotting and shows specific reactivity by with ESRP2 transfected cell lysates..  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 77.4 kDa.

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:1000;

Type: Primary
Antigen: ESRP2
Clonality: Monoclonal
Clone: 22C5.H7.A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge