Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-KDELR1 Mouse Monoclonal Antibody (Unconjugated) [clone: KR-10]

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

500-301-F86
ROCK500-301-F86EA 841 EUR InStock
ROCK500-301-F86
Anti-KDELR1 Mouse Monoclonal Antibody (Unconjugated) [clone: KR-10]
Antikörper
Anti-KDEL Antibody is suitable for WB, IP, IF microscopy and IHC. Expect a band approximately ~25kDa Protein corresponding to the molecular mass of KDEL on SDS-PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: KDELR1
Clonality: Monoclonal
Clone: KR-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat, Mouse, Hamster, Rabbit, Pig, Sheep, Dog, Chicken, Drosophilia, Xenopus, Bovine

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge