Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-VDAC1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-882S
ROCK600-401-882SEA 226 EUR InStock
ROCK600-401-882S
Anti-VDAC1 Rabbit Polyclonal Antibody
Antikörper
VDAC/Porin Antibody has been tested for use in ELISA and western blot. Specific conditions for reactivity should be optimised by the end user. Expect a band at ~30 to 33 kDa in size.

Recommended dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:500 - 1:2000; Immunochemistry: Yes; other dilution: User Optimised

Type: Primary
Antigen: VDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Human, Rabbit, Bovine, Pig, Goat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge