Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-LACl Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-B04
ROCK600-401-B04EA 821 EUR InStock
ROCK600-401-B04
Anti-LACl Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested for use in ELISA and western blotting using recombinant Lac I Protein.   Specific conditions for reactivity and detection of Lac I should be optimized by the end user. Expect a band approximately ~38 kDa.

Recommended Dilutions: ELISA: 1:90,000 – 1:100000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: LACl
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli, S.Sonnei, E. coli

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge