Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-IL34 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-C06
ROCK600-401-C06EA 944 EUR InStock
ROCK600-401-C06
Anti-IL34 Rabbit Polyclonal Antibody
Antikörper
Anti-IL-34 Antibody has been tested for use in ELISA and WB. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~48 kDa in size corresponding to IL-34 by Western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 0.25 - 0.5 µg/ml;

Type: Primary
Antigen: IL34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge