Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TH Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-C99
ROCK600-401-C99EA 976 EUR InStock
ROCK600-401-C99
Anti-TH Rabbit Polyclonal Antibody
Antikörper
Anti-Tyrosine Hydroxylase pS40 antibody is suitable for use in ELISA, Western Blotting, IHC and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 60 kDa in size corresponding to the tyrosine hydroxylase.

Recommended Dilutions: ELISA: 1:10000; Immunofluorescence: 1:1000; Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge