Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLRA1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

600-401-D65
ROCK600-401-D65EA 976 EUR InStock
ROCK600-401-D65
Anti-GLRA1 Rabbit Polyclonal Antibody
Antikörper
Anti-Glycine Receptor (Rabbit) antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 48 kDa in size corresponding to the alpha 1 and alpha 2 subunit of the GABA(A) Receptor.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000
0.05 M Tris Chloride, 0.15 M Sodium Chloride, pH 7.4;

Type: Primary
Antigen: GLRA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse, Human

Order Now
 

PRODUKT

TECHNISCHE DATEN

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge